missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium bromate, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 446945000

 View more versions of this product

Catalog No. AC446945000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium bromate
7758-01-2
BrKO3
15,7738
Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water
[O-]Br(=O)=O.[K+]
167.01
CHEBI:38211
ACS Reagent
3.27
350.0°C
500 g
KBrO3
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium;bromate
23673461
167.01
Plastic bottle
3.2700g/mL

Chemical Identifiers

7758-01-2
167.01
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
CHEBI:38211
[O-]Br(=O)=O.[K+]
BrKO3
OCATYIAKPYKMPG-UHFFFAOYSA-M
23673461
potassium;bromate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 231-829-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only