missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium bromate, ACS reagent, Thermo Scientific Chemicals
$133.84 - $311.81
Chemical Identifiers
CAS | 7758-01-2 |
---|---|
Molecular Formula | BrKO3 |
Molecular Weight (g/mol) | 167.01 |
InChI Key | OCATYIAKPYKMPG-UHFFFAOYSA-M |
Synonym | potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat |
PubChem CID | 23673461 |
ChEBI | CHEBI:38211 |
IUPAC Name | potassium;bromate |
SMILES | [O-]Br(=O)=O.[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC446941000
|
Thermo Scientific Chemicals
446941000 |
100 g | Plastic bottle |
Each for $133.84
|
|
||||
AC446945000
|
Thermo Scientific Chemicals
446945000 |
500 g | Plastic bottle |
Each for $311.81
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-01-2 | |
167.01 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
CHEBI:38211 | |
[O-]Br(=O)=O.[K+] |
BrKO3 | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
23673461 | |
potassium;bromate |
Specifications
350.0°C | |
100 g | |
KBrO3 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
potassium;bromate | |
23673461 | |
167.01 | |
Plastic bottle | |
3.2700g/mL |
7758-01-2 | |
BrKO3 | |
15,7738 | |
Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water | |
[O-]Br(=O)=O.[K+] | |
167.01 | |
CHEBI:38211 | |
ACS Reagent | |
3.27 | |
Potassium bromate |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 231-829-8
RUO – Research Use Only