Learn More
Potassium carbonate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 424080025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentSpecifications
Potassium carbonate | |
584-08-7 | |
Fine Granular Powder | |
99+% | |
K2CO3 | |
01,1014; 04,50; 05,552; 08,408; 09,382; 10,323; 11,433; 12,403; 13,233; 15,167 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
dipotassium;carbonate | |
11430 | |
≥99% | |
Plastic bottle |
891.0°C | |
White | |
2.5 kg | |
CK2O3 | |
MFCD00011382 | |
15, 7740 | |
Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone | |
[K+].[K+].[O-]C([O-])=O | |
138.21 | |
138.21 | |
ACS Reagent |
Chemical Identifiers
584-08-7 | |
138.21 | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
11430 |
CK2O3 | |
MFCD00011382 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
[K+].[K+].[O-]C([O-])=O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If skin irri
GHS Signal Word: Warning
EINECSNumber : 209-529-3
RTECSNumber : TS7750000
TSCA : TSCA
RUO – Research Use Only