Learn More
Potassium carbonate, 99+%, ACS reagent, anhydrous
CAS: 584-08-7 | CK2O3 | 138.21 g/mol
$72.86 - $2065.12
Chemical Identifiers
| CAS | 584-08-7 |
|---|---|
| Molecular Formula | CK2O3 |
| Molecular Weight (g/mol) | 138.21 |
| MDL Number | MFCD00011382 |
| InChI Key | BWHMMNNQKKPAPP-UHFFFAOYSA-L |
| Synonym | potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate |
| PubChem CID | 11430 |
| SMILES | [K+].[K+].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC424081000
|
Thermo Scientific Chemicals
424081000 |
100 g | Plastic bottle |
Each for $72.86
|
|
||||
|
AC424085000
|
Thermo Scientific Chemicals
424085000 |
500 g | Plastic Bottle |
Each for $213.56
|
|
||||
|
AC424080025
|
Thermo Scientific Chemicals
424080025 |
2.5 kg | Plastic bottle |
Each for $640.68
|
|
||||
|
AC424080100
|
Thermo Scientific Chemicals
424080100 |
10 kg | Plastic Drum |
Each for $2,065.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentChemical Identifiers
| 584-08-7 | |
| 138.21 | |
| BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
| 11430 |
| CK2O3 | |
| MFCD00011382 | |
| potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
| [K+].[K+].[O-]C([O-])=O |
Specifications
| 891.0°C | |
| White | |
| 100 g | |
| CK2O3 | |
| MFCD00011382 | |
| 15, 7740 | |
| Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone | |
| [K+].[K+].[O-]C([O-])=O | |
| 138.21 | |
| 138.21 | |
| ACS Reagent | |
| Potassium carbonate |
| 584-08-7 | |
| Fine Granular Powder | |
| 99+% | |
| K2CO3 | |
| 01,1014; 04,50; 05,552; 08,408; 09,382; 10,323; 11,433; 12,403; 13,233; 15,167 | |
| potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
| BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
| dipotassium;carbonate | |
| 11430 | |
| ≥99% | |
| Plastic bottle |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If skin irri
GHS Signal Word: Warning
EINECSNumber : 209-529-3
RTECSNumber : TS7750000
TSCA : TSCA
RUO – Research Use Only