missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium carbonate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424085000

 View more versions of this product

Catalog No. AC424085000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications

Specifications

Potassium Carbonate
584-08-7
Fine Granular Powder
99+%
K2CO3
01,1014; 04,50; 05,552; 08,408; 09,382; 10,323; 11,433; 12,403; 13,233; 15,167
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium carbonate
11430
≥99%
Plastic Bottle
891.0°C
White
500 g
CK2O3
MFCD00011382
15, 7740
Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone
[K+].[K+].[O-]C([O-])=O
138.21
138.21
ACS Reagent

Chemical Identifiers

584-08-7
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
[K+].[K+].[O-]C([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

Warning

EINECSNumber : 209-529-3

RTECSNumber : TS7750000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only