missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium cyanide, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1327336

 View more versions of this product

Catalog No. AAL1327336


Explore available promotions

Description

Description

Potassium cyanide is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium cyanide is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.

Solubility
Highly soluble in water. Soluble in methanol, glycerol, and formamide. Slightly soluble in ethanol.

Notes
Deliquescent solid. Store in airtight containers. Incompatible with acids.
Specifications

Specifications

1625°C
634°C
500 g
KCN
UN1680
14,7626
Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol.
[C-]#N.[K+]
65.116
65.12
Hygroscopic
Potassium cyanide
151-50-8
CKN
MFCD00011397
4652394
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium;cyanide
9032
≥97%
1.52

Chemical Identifiers

151-50-8
65.116
NNFCIKHAZHQZJG-UHFFFAOYSA-N
9032
[C-]#N.[K+]
CKN
MFCD00011397
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
potassium;cyanide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P390-P501c

H290-H300+H310+H330-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: POTASSIUM CYANIDE, SOLID

EINECSNumber : 205-792-3

RTECSNumber : TS8750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only