Learn More
Potassium cyanide, 97+%, Thermo Scientific Chemicals
$88.55 - $181.52
Chemical Identifiers
CAS | 151-50-8 |
---|---|
Molecular Formula | CKN |
Molecular Weight (g/mol) | 65.116 |
MDL Number | MFCD00011397 |
InChI Key | NNFCIKHAZHQZJG-UHFFFAOYSA-N |
Synonym | potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french |
PubChem CID | 9032 |
IUPAC Name | potassium;cyanide |
SMILES | [C-]#N.[K+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1327322
|
Thermo Scientific Chemicals
L1327322 |
100 g |
Each for $88.55
|
|
|||||
AAL1327336
|
Thermo Scientific Chemicals
L1327336 |
500 g |
Each for $181.52
|
|
|||||
Description
Potassium cyanide is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium cyanide is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.
Solubility
Highly soluble in water. Soluble in methanol, glycerol, and formamide. Slightly soluble in ethanol.
Notes
Deliquescent solid. Store in airtight containers. Incompatible with acids.
Chemical Identifiers
151-50-8 | |
65.116 | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
9032 | |
[C-]#N.[K+] |
CKN | |
MFCD00011397 | |
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french | |
potassium;cyanide |
Specifications
1625°C | |
151-50-8 | |
CKN | |
MFCD00011397 | |
4652394 | |
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
potassium;cyanide | |
9032 | |
≥97% | |
1.52 |
634°C | |
100 g | |
KCN | |
UN1680 | |
14,7626 | |
Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol. | |
[C-]#N.[K+] | |
65.116 | |
65.12 | |
Hygroscopic | |
Potassium cyanide |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P390-P501c
H290-H300+H310+H330-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: POTASSIUM CYANIDE, SOLID
EINECSNumber : 205-792-3
RTECSNumber : TS8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only