missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Fluoride, Anhydrous, Purified, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P162125KG

 View more versions of this product

Catalog No. 18612641


Explore available promotions

Description

Description

Specifications

Specifications

7789-23-3
2.5 kg
MFCD00011398
[F-].[K+]
58.10
Purified
1
FK
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
99%
Poly Pail

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
[F-].[K+]
FK
MFCD00011398
potassium fluoride
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.