missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Fluoride, Anhydrous, Purified, 99%, Spectrum™ Chemical
$142.29 - $11039.85
Chemical Identifiers
CAS | 7789-23-3 |
---|---|
Molecular Formula | FK |
Molecular Weight (g/mol) | 58.10 |
MDL Number | MFCD00011398 |
InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
IUPAC Name | potassium fluoride |
SMILES | [F-].[K+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18612638
|
Spectrum Chemical Mfg Cor
P162125GM |
25 g | Poly Bottle |
Each for $142.29
|
|
||||
18612639
|
Spectrum Chemical Mfg Cor
P1621125GM |
125 g | Poly Bottle |
Each for $208.41
|
|
||||
18612640
|
Spectrum Chemical Mfg Cor
P1621500GM |
500 g | Poly Bottle |
Each for $499.65
|
|
||||
18612641
|
Spectrum Chemical Mfg Cor
P162125KG |
2.5 kg | Poly Pail |
Each for $1,339.44
|
|
||||
18612642
|
Spectrum Chemical Mfg Cor
P162112KG |
12 kg | Fiber Drum |
Each for $4,099.35
|
|
||||
18612643
|
Spectrum Chemical Mfg Cor
P162145KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7789-23-3 | |
58.10 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
[F-].[K+] |
FK | |
MFCD00011398 | |
potassium fluoride |
Specifications
7789-23-3 | |
25 g | |
MFCD00011398 | |
[F-].[K+] | |
58.10 | |
Purified |
1 | |
FK | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
potassium fluoride | |
99% | |
Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.