missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%

Maximum level of impurities: Insoluble matter 0.01%, SO4 0.005%, Fe 0.001%, Cl 0.02%, Heavy Metals (as Pb) 0.001% | CAS: 13455-24-8 | HI2KO6 | 389.909 g/mol

$132.40 - $367.18

Chemical Identifiers

CAS 13455-24-8
Molecular Formula HI2KO6
Molecular Weight (g/mol) 389.909
MDL Number MFCD00011400
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
PubChem CID 23700942
IUPAC Name potassium;iodic acid;iodate
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4198614
View Documents
Thermo Scientific Chemicals
04198614
25 g
Each for $132.40
Only null left
 
AA4198622
View Documents
Thermo Scientific Chemicals
04198622
100 g
Each for $367.18
Only null left
 
Description

Description

Potassium hydrogen iodate acts as an oxidizing agent. Further, it serves as an intermediate for the synthesis of potassium iodate and also used as an analysis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13455-24-8
389.909
ACAYDTMSDROWHW-UHFFFAOYSA-M
potassium;iodic acid;iodate
HI2KO6
MFCD00011400
23700942
OI(=O)=O.[O-]I(=O)=O.[K+]

Specifications

13455-24-8
Powder
MFCD00011400
Soluble in water
OI(=O)=O.[O-]I(=O)=O.[K+]
389.909
23700942
ACS
99.95-100.05%
HI2KO6
White
ACAYDTMSDROWHW-UHFFFAOYSA-M
potassium;iodic acid;iodate
25 g
389.91
Potassium hydrogen iodate, Primary Standard
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 236-650-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only