Learn More
Potassium hydroxide, 99.98%, (trace metal basis), pellets, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 437131000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
Potassium hydroxide | |
1320.0°C | |
360.0°C | |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
100 g | |
[OH-].[K+] | |
56.11 | |
99.98% | |
Plastic bottle |
White | |
1310-58-3 | |
99.98% | |
KOH | |
15,7761 | |
Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C),soluble in ethanol (400g/L),soluble in ammonia,slightly soluble in ether | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
56.11 | |
Trace Metal Basis |
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
[OH-].[K+] |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 215-181-3
RUO – Research Use Only