missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium hydroxide, 99.98%, (trace metal basis), pellets, Thermo Scientific Chemicals

$183.44 - $539.46

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC437131000
SDS View Documents
Thermo Scientific Chemicals
437131000
100 g Plastic bottle
Each for $183.44
 
AC437135000
SDS View Documents
Thermo Scientific Chemicals
437135000
500 g Plastic bottle
Each for $539.46
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]

Specifications

White
1310-58-3
99.98%
KOH
15,7761
Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C),soluble in ethanol (400g/L),soluble in ammonia,slightly soluble in ether
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
56.11
Trace Metal Basis
Potassium hydroxide
1320.0°C
360.0°C
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
100 g
[OH-].[K+]
56.11
99.98%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 215-181-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only