missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Hydroxide Solution, 0.5N (N/2) (In Methanol/Certified), Fisher Chemical™
Titrated at 25°C to pH 8.2 (phenolphthalein endpoint) against standardized HCl
Supplier: Thermo Fisher Scientific FLSP2221
Specifications
Potassium Hydroxide Solution | |
0.5N (N/2), in methanol | |
1310-58-3,67-56-1,7732-18-5 | |
HKO | |
0.497 to 0.500N | |
0.8g/cm³ | |
1 L | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Certified | |
Pass Test | |
Liquid |
Colorless | |
65.6°C | |
-100°C | |
MFCD00003553 | |
2924 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium hydroxide | |
14797 | |
56.11 | |
0.5N (N/2) | |
Glass Bottle |
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
potassium hydroxide |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 | |
[OH-].[K+] |
Safety and Handling
DOTInformation : DOT Class 3, : Flammable Liquid