missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Iodide, USP, 99-101.5%, Spectrum™ Chemical
PO185, 7681-11-0, KI
Supplier: Spectrum Chemical Mfg Cor PO185500GM
Description
Spectrum™ Chemical Potassium Iodide, USP is used to treat hyperthyroidism in radiation emergencies and to protect the thyroid gland when certain types of radiopharmaceuticals are used. All Spectrum Chemical USP products, including all USP potassium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
7681-11-0 | |
500 g | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium iodide | |
99 to 101.5% | |
0.01 |
1 | |
IK | |
[K+].[I-] | |
166.00 | |
USP | |
Amber Glass Bottle |
Chemical Identifiers
7681-11-0 | |
166.00 | |
potassium iodide |
IK | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
[K+].[I-] |