missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Iodide, USP, 99-101.5%, Spectrum™ Chemical
PO185, 7681-11-0, KI
$416.47 - $32519.44
Chemical Identifiers
CAS | 7681-11-0 |
---|---|
Molecular Formula | IK |
Molecular Weight (g/mol) | 166.00 |
InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
IUPAC Name | potassium iodide |
SMILES | [K+].[I-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18605632
|
Spectrum Chemical Mfg Cor
PO185125GM |
125 g | Amber Glass Bottle |
Each for $416.47
|
|
||||
18605633
|
Spectrum Chemical Mfg Cor
PO185500GM |
500 g | Amber Glass Bottle |
Each for $1,439.62
|
|
||||
18605635
|
Spectrum Chemical Mfg Cor
PO18525KGGL |
2.5 kg | Amber Glass Bottle |
Each for $5,029.82
|
|
||||
18605638
|
Spectrum Chemical Mfg Cor
PO18525KGPL |
2.5 kg | Poly Bottle |
Each for $4,921.82
|
|
||||
18605634
|
Spectrum Chemical Mfg Cor
PO185500GMCS6 |
6 x 500 g | Amber Glass Bottle |
Case of 6 Each for $6,400.21
|
|
||||
18605636
|
Spectrum Chemical Mfg Cor
PO18525KGGLCS4 |
4 x 2.5 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18605639
|
Spectrum Chemical Mfg Cor
PO18525KGPLCS4 |
4 x 2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18605637
|
Spectrum Chemical Mfg Cor
PO18512KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18605640
|
Spectrum Chemical Mfg Cor
PO18545KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Potassium Iodide, USP is used to treat hyperthyroidism in radiation emergencies and to protect the thyroid gland when certain types of radiopharmaceuticals are used. All Spectrum Chemical USP products, including all USP potassium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
7681-11-0 | |
166.00 | |
potassium iodide |
IK | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
[K+].[I-] |
Specifications
7681-11-0 | |
125 g | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium iodide | |
99 to 101.5% | |
0.01 |
1 | |
IK | |
[K+].[I-] | |
166.00 | |
USP | |
Amber Glass Bottle |