missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Iodide, USP, 99-101.5%, Spectrum™ Chemical

PO185, 7681-11-0, KI

Supplier:  Spectrum Chemical Mfg Cor PO185500GMCS6

 View more versions of this product

Catalog No. 18605634


Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Iodide, USP is used to treat hyperthyroidism in radiation emergencies and to protect the thyroid gland when certain types of radiopharmaceuticals are used. All Spectrum Chemical USP products, including all USP potassium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7681-11-0
6 x 500 g
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide
99 to 101.5%
Amber Glass Bottle
1
IK
[K+].[I-]
166.00
0.01

Chemical Identifiers

7681-11-0
166.00
potassium iodide
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
[K+].[I-]
SDS
Documents

Documents

Promotions

Promotions