missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 418275000

 View more versions of this product

Catalog No. AC418275000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Potassium metabisulfite
97%
White to Yellow
500 g
K2O5S2
MFCD00167605
potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
RWPGFSMJFRPDDP-UHFFFAOYSA-L
dipotassium sulfinatosulfonate
28019
97%
Glass bottle
150.0°C
16731-55-8
Fine Crystalline Powder
96% min. (Iodimetry)
K2S2O5
15, 7766
Solubility in water: 45g/L (20°C). Other solubilities: insoluble in ethanol
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
222.31
222.33
Extra Pure

Chemical Identifiers

16731-55-8
222.31
RWPGFSMJFRPDDP-UHFFFAOYSA-L
28019
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
K2O5S2
MFCD00167605
potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
dipotassium sulfinatosulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Contact with acids liberates toxic gas.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 240-795-3

RTECSNumber : TT4920000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only