missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium metabisulfite, 97%, extra pure, Thermo Scientific Chemicals

$67.67 - $307.71

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
PubChem CID 28019
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC418270250
SDS View Documents
Thermo Scientific Chemicals
418270250
25 g Glass bottle
Each for $67.67
 
AC418275000
SDS View Documents
Thermo Scientific Chemicals
418275000
500 g Glass bottle
Each for $92.16
 
AC418270025
SDS View Documents
Thermo Scientific Chemicals
418270025
2.5 kg Plastic bottle
Each for $307.71
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

16731-55-8
222.31
RWPGFSMJFRPDDP-UHFFFAOYSA-L
28019
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
K2O5S2
MFCD00167605
potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
dipotassium sulfinatosulfonate

Specifications

150.0°C
White to Yellow
25 g
K2O5S2
MFCD00167605
potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
RWPGFSMJFRPDDP-UHFFFAOYSA-L
dipotassium sulfinatosulfonate
28019
97%
Glass bottle
16731-55-8
Fine Crystalline Powder
96% min. (Iodimetry)
K2S2O5
15, 7766
Solubility in water: 45g/L (20°C). Other solubilities: insoluble in ethanol
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
222.31
222.33
Extra Pure
Potassium metabisulfite, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Contact with acids liberates toxic gas.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 240-795-3

RTECSNumber : TT4920000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only