missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic Anhydrous (Crystalline Powder/Certified ACS), Fisher Chemical™

$271.25 - $15970.58

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
P288-100
SDS View Documents Promotion Details
Thermo Fisher Scientific
P288100
100 g Plastic Bottle
Each for $271.25
Case of 6 Each for $1,627.50
Only null left
 
P288-500 Edge
SDS View Documents Promotion Details
Thermo Fisher Scientific
P288500
500 g Plastic Bottle
Each for $353.08
Case of 6 Each for $2,118.48
Only null left
Edge
 
P288-3
SDS View Documents Promotion Details
Thermo Fisher Scientific
P2883
3 kg Plastic Bottle
Each for $1,178.33
Case of 4 Each for $4,713.32
Only null left
 
P288-50
SDS View Documents Promotion Details
Thermo Fisher Scientific
P28850
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

11-4-7758
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
24450
dipotassium;hydrogen phosphate
HK2O4P
MFCD00011383
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
CHEBI:32031
OP(=O)([O-])[O-].[K+].[K+]

Specifications

340°C
White
Solid
≥98.0 %
MFCD00011383
Soluble in water
OP(=O)([O-])[O-].[K+].[K+]
174.17
CHEBI:32031
≥98.0%
Pass Test
Plastic Bottle
Potassium Phosphate Dibasic Anhydrous
11-4-7758
8.5 to 9.6
100 g
HK2O4P
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium;hydrogen phosphate
24450
174.18
Certified ACS
1% max.
>180°C
Videos
Safety and Handling

Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0

CAUTION!

SDS
missing translation for 'documents'

missing translation for 'documents'