missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Phosphate Dibasic Anhydrous (Crystalline Powder/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific P288100
Specifications
Potassium Phosphate Dibasic Anhydrous | |
7758-11-4 | |
8.5 to 9.6 | |
100 g | |
HK2O4P | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate | |
24450 | |
174.18 | |
Certified ACS | |
1% max. | |
>180°C |
340°C | |
White | |
Solid | |
≥98.0 % | |
MFCD00011383 | |
Soluble in water | |
OP(=O)([O-])[O-].[K+].[K+] | |
174.17 | |
CHEBI:32031 | |
≥98.0% | |
Pass Test | |
Plastic Bottle |
Chemical Identifiers
7758-11-4 | |
174.17 | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
24450 | |
dipotassium;hydrogen phosphate |
HK2O4P | |
MFCD00011383 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
CHEBI:32031 | |
OP(=O)([O-])[O-].[K+].[K+] |
Safety and Handling
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:0
CAUTION!