missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium phosphate, monobasic, 99+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424200025

 View more versions of this product

Catalog No. AC424200025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium phosphate, monobasic
7778-77-0
4.1 to 4.5 (5% soln. at 25°C)
2.5 kg
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium;dihydrogen phosphate
516951
136.09
ACS Reagent
Plastic bottle
253.0°C
Colorless to White
Crystals
99+%
KH2PO4
15, 7780
Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
≥99%
0.2% max. (2 g, 105°C, 2 hrs)
>250°C

Chemical Identifiers

7778-77-0
136.08
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
[K+].OP(O)([O-])=O
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
CHEBI:63036
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 231-913-4

RTECSNumber : TC6615500

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only