missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate, monobasic, 99+%, ACS reagent
CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
$113.07 - $486.12
Chemical Identifiers
| CAS | 7778-77-0 |
|---|---|
| Molecular Formula | H2KO4P |
| Molecular Weight (g/mol) | 136.08 |
| MDL Number | MFCD00011401 MFCD00147253 |
| InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
| Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
| PubChem CID | 516951 |
| ChEBI | CHEBI:63036 |
| SMILES | [K+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC424200250
|
Thermo Scientific Chemicals
424200250 |
25 g | Plastic bottle |
Each for $113.07
|
|
||||
|
AC424205000
|
Thermo Scientific Chemicals
424205000 |
500 g | Plastic bottle |
Each for $172.21
|
|
||||
|
AC424200025
|
Thermo Scientific Chemicals
424200025 |
2.5 kg | Plastic bottle |
Each for $486.12
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| 516951 | |
| [K+].OP(O)([O-])=O |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| CHEBI:63036 |
Specifications
| 253.0°C | |
| Colorless to White | |
| Crystals | |
| 99+% | |
| KH2PO4 | |
| 15, 7780 | |
| Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| CHEBI:63036 | |
| ≥99% | |
| 0.2% max. (2 g, 105°C, 2 hrs) | |
| >250°C |
| 7778-77-0 | |
| 4.1 to 4.5 (5% soln. at 25°C) | |
| 25 g | |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium dihydrogen phosphate | |
| 516951 | |
| 136.09 | |
| ACS Reagent | |
| Plastic bottle | |
| Potassium phosphate, monobasic |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 231-913-4
RTECSNumber : TC6615500
TSCA : TSCA
RUO – Research Use Only