missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium selenite, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03943614

 View more versions of this product

Catalog No. AA3943614


Explore available promotions

Description

Description

One among the compounds used in cattle feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium selenite
10431-47-7
Powder
K2O3Se
UN2630
Soluble in ethyl alcohol,water.
[O-][Se](=O)[O-].[K+].[K+]
205.165
205.16
2.851 g/mL
875°C (decomposition)
White
25 g
MFCD00049663
potassium selenite, dipotassium selenite, ccris 6885, selenious acid, dipotassium salt, kaliumselenat, acmc-20ajua, selenious acid, potassium salt 1:2, dipotassium ion selenit, selenious acid,potassium salt 1:2
RNGFNLJMTFPHBS-UHFFFAOYSA-L
dipotassium;selenite
150192
Hygroscopic

Chemical Identifiers

10431-47-7
205.165
RNGFNLJMTFPHBS-UHFFFAOYSA-L
150192
[O-][Se](=O)[O-].[K+].[K+]
K2O3Se
MFCD00049663
potassium selenite, dipotassium selenite, ccris 6885, selenious acid, dipotassium salt, kaliumselenat, acmc-20ajua, selenious acid, potassium salt 1:2, dipotassium ion selenit, selenious acid,potassium salt 1:2
dipotassium;selenite
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES

EINECSNumber : 233-909-8

RTECSNumber : VS7200000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only