Learn More
Potassium selenite, Thermo Scientific Chemicals
$211.44 - $717.48
Chemical Identifiers
CAS | 10431-47-7 |
---|---|
Molecular Formula | K2O3Se |
Molecular Weight (g/mol) | 205.165 |
MDL Number | MFCD00049663 |
InChI Key | RNGFNLJMTFPHBS-UHFFFAOYSA-L |
Synonym | potassium selenite, dipotassium selenite, ccris 6885, selenious acid, dipotassium salt, kaliumselenat, acmc-20ajua, selenious acid, potassium salt 1:2, dipotassium ion selenit, selenious acid,potassium salt 1:2 |
PubChem CID | 150192 |
IUPAC Name | dipotassium;selenite |
SMILES | [O-][Se](=O)[O-].[K+].[K+] |
Description
One among the compounds used in cattle feeds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10431-47-7 | |
205.165 | |
RNGFNLJMTFPHBS-UHFFFAOYSA-L | |
150192 | |
[O-][Se](=O)[O-].[K+].[K+] |
K2O3Se | |
MFCD00049663 | |
potassium selenite, dipotassium selenite, ccris 6885, selenious acid, dipotassium salt, kaliumselenat, acmc-20ajua, selenious acid, potassium salt 1:2, dipotassium ion selenit, selenious acid,potassium salt 1:2 | |
dipotassium;selenite |
Specifications
875°C (decomposition) | |
White | |
25 g | |
MFCD00049663 | |
potassium selenite, dipotassium selenite, ccris 6885, selenious acid, dipotassium salt, kaliumselenat, acmc-20ajua, selenious acid, potassium salt 1:2, dipotassium ion selenit, selenious acid,potassium salt 1:2 | |
RNGFNLJMTFPHBS-UHFFFAOYSA-L | |
dipotassium;selenite | |
150192 | |
Hygroscopic | |
Potassium selenite |
10431-47-7 | |
Powder | |
K2O3Se | |
UN2630 | |
Soluble in ethyl alcohol,water. | |
[O-][Se](=O)[O-].[K+].[K+] | |
205.165 | |
205.16 | |
2.851 g/mL |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
EINECSNumber : 233-909-8
RTECSNumber : VS7200000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only