missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium sorbate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A128440I

 View more versions of this product

Catalog No. AAA128440I


Explore available promotions

Description

Description

Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.

Solubility
Soluble in water, alcohol, propylene glycol. Slightly soluble in acetoneand chloroform, corn oil and ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Potassium sorbate
>300°C
8.3
C6H7KO2
MFCD00016546
14,7671
Soluble in water,alcohol,propylene glycol. Slightly soluble in acetoneand chloroform,corn oil and ether. Insoluble in benzene.
CC=CC=CC(=O)[O-].[K+]
150.218
CHEBI:77868
99%
24634-61-5
1.361
99%
CH3CH=CH-CH=CHCO2K
5000 g
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
CHHHXKFHOYLYRE-STWYSWDKSA-M
potassium;(2E,4E)-hexa-2,4-dienoate
23676745
150.22

Chemical Identifiers

24634-61-5
150.218
CHHHXKFHOYLYRE-STWYSWDKSA-M
23676745
potassium;(2E,4E)-hexa-2,4-dienoate
C6H7KO2
MFCD00016546
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
CHEBI:77868
CC=CC=CC(=O)[O-].[K+]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 246-376-1

RTECSNumber : WG2170000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only