missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium sorbate, 99%, Thermo Scientific Chemicals

$82.63 - $878.16

Chemical Identifiers

CAS 24634-61-5
Molecular Formula C6H7KO2
Molecular Weight (g/mol) 150.218
MDL Number MFCD00016546
InChI Key CHHHXKFHOYLYRE-STWYSWDKSA-M
Synonym potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
PubChem CID 23676745
ChEBI CHEBI:77868
IUPAC Name potassium;(2E,4E)-hexa-2,4-dienoate
SMILES CC=CC=CC(=O)[O-].[K+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1284430
View Documents
Thermo Scientific Chemicals
A1284430
250 g
Each for $82.63
 
AAA128440B
View Documents
Thermo Scientific Chemicals
A128440B
1000 g
Each for $254.14
 
AAA128440I
View Documents
Thermo Scientific Chemicals
A128440I
5000 g
Each for $878.16
 
Description

Description

Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.

Solubility
Soluble in water, alcohol, propylene glycol. Slightly soluble in acetoneand chloroform, corn oil and ether. Insoluble in benzene.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

24634-61-5
150.218
CHHHXKFHOYLYRE-STWYSWDKSA-M
23676745
potassium;(2E,4E)-hexa-2,4-dienoate
C6H7KO2
MFCD00016546
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
CHEBI:77868
CC=CC=CC(=O)[O-].[K+]

Specifications

24634-61-5
1.361
C6H7KO2
MFCD00016546
14,7671
Soluble in water,alcohol,propylene glycol. Slightly soluble in acetoneand chloroform,corn oil and ether. Insoluble in benzene.
CC=CC=CC(=O)[O-].[K+]
150.218
CHEBI:77868
99%
>300°C
99%
CH3CH=CH-CH=CHCO2K
250 g
potassium sorbate, sorbistat-k, potassium 2,4-hexadienoate, sorbistat potassium, potassium e,e-sorbate, sorbic acid potassium salt, potassium 2e,4e-hexa-2,4-dienoate, bb powder, sorbistat-potassium, caswell no. 701c
CHHHXKFHOYLYRE-STWYSWDKSA-M
potassium;(2E,4E)-hexa-2,4-dienoate
23676745
150.22
Potassium Sorbate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 246-376-1

RTECSNumber : WG2170000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only