missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tetraborate tetrahydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03943536

 View more versions of this product

Catalog No. AA3943536


Explore available promotions

Description

Description

It is an excellent solvent for metallic oxides at high temperatures. It is a good buffering agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium tetraborate tetrahydrate
White
Granules
99%
MFCD00149924
Soluble in water
Odorless
1.74 g/cm3 at 20°C
(decomposition)
12045-78-2
500 g
K2B4O7+4H2O
Potassium borate (tetra)
305.51 (233.44 Anhydrous)
White crystals

Chemical Identifiers

12045-78-2
K2B4O7+4H2O
MFCD00149924
Potassium borate (tetra)
Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H361d

EINECSNumber : 215-575-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only