missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium tetraborate tetrahydrate, 99%, Thermo Scientific Chemicals

$83.11 - $227.47

Chemical Identifiers

CAS 12045-78-2
Molecular Formula K2B4O7+4H2O
MDL Number MFCD00149924
Synonym Potassium borate (tetra)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3943536
View Documents
Thermo Scientific Chemicals
03943536
500 g
Each for $83.11
 
AA39435A3
View Documents
Thermo Scientific Chemicals
039435A3
2 kg
Each for $227.47
 
Description

Description

It is an excellent solvent for metallic oxides at high temperatures. It is a good buffering agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12045-78-2
K2B4O7+4H2O
MFCD00149924
Potassium borate (tetra)

Specifications

(decomposition)
12045-78-2
500 g
K2B4O7+4H2O
Potassium borate (tetra)
305.51 (233.44 Anhydrous)
White crystals
Potassium tetraborate tetrahydrate
White
Granules
99%
MFCD00149924
Soluble in water
Odorless
1.74 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H361d

EINECSNumber : 215-575-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only