missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium thiosulfate hydrate, 96+%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 388691000

 View more versions of this product

Catalog No. AC388691000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Potassium thiosulfate hydrate
1264198-56-2
Liquid
96.0 to 104% (On dry substance) (Iodimetry)
K2S2O3·xH2O
Solubility in water: soluble. Other solubilities: insoluble in alcohol
[K+].[K+].[O-]S([S-])(=O)=O
190.31
190.33
Extra Pure
Plastic bottle
Colorless to Yellow
6 to 9 (5% soln.)
100 g
K2O3S2
MFCD00150405
FGRVOLIFQGXPCT-UHFFFAOYSA-L
dipotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
61501
≥96%
4% max. (150°C)

Chemical Identifiers

1264198-56-2
190.31
FGRVOLIFQGXPCT-UHFFFAOYSA-L
[K+].[K+].[O-]S([S-])(=O)=O
K2O3S2
MFCD00150405
61501
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only