Learn More
Potassium thiosulfate hydrate, 96+%, extra pure, Thermo Scientific Chemicals
$271.30 - $669.39
Chemical Identifiers
CAS | 1264198-56-2 |
---|---|
Molecular Formula | K2O3S2 |
Molecular Weight (g/mol) | 190.31 |
MDL Number | MFCD00150405 |
InChI Key | FGRVOLIFQGXPCT-UHFFFAOYSA-L |
PubChem CID | 61501 |
SMILES | [K+].[K+].[O-]S([S-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC388691000
|
Thermo Scientific Chemicals
388691000 |
100 g | Plastic bottle |
Each for $271.30
|
|
||||
AC388695000
|
Thermo Scientific Chemicals
388695000 |
500 g | Plastic bottle |
Each for $669.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
1264198-56-2 | |
190.31 | |
FGRVOLIFQGXPCT-UHFFFAOYSA-L | |
[K+].[K+].[O-]S([S-])(=O)=O |
K2O3S2 | |
MFCD00150405 | |
61501 |
Specifications
Colorless to Yellow | |
6 to 9 (5% soln.) | |
100 g | |
K2O3S2 | |
MFCD00150405 | |
FGRVOLIFQGXPCT-UHFFFAOYSA-L | |
dipotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
61501 | |
≥96% | |
4% max. (150°C) | |
Potassium thiosulfate hydrate |
1264198-56-2 | |
Liquid | |
96.0 to 104% (On dry substance) (Iodimetry) | |
K2S2O3·xH2O | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
[K+].[K+].[O-]S([S-])(=O)=O | |
190.31 | |
190.33 | |
Extra Pure | |
Plastic bottle |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only