missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Praseodymium(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03566309

 View more versions of this product

Catalog No. AA3566309


Explore available promotions

Description

Description

Praseodymium(III) oxide is used as a pigment in glass, optic and ceramic industries. Praseodymium-doped glass called didymium glass is used in welding goggles, since it blocks infra-red radiation. Further, it is used as a dielectric in combination with silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Praseodymium(III) oxide
12036-32-7
10 g
O3Pr2
14,7712
Insoluble in water. Moderately soluble in strong mineral acids.
[O-2].[O-2].[O-2].[Pr+3].[Pr+3]
329.812
329.83
Odorless
2300°C
Sintered Lump
(metals basis)
MFCD00011178
dipraseodymium trioxide, praseodymium iii oxide, praseodymium oxide pr2o3, praseodynium oxide, dipraseodymium 3+ ion trioxidandiide, praseodymium iii oxide trace metals basis 10g
MMKQUGHLEMYQSG-UHFFFAOYSA-N
oxygen(2-);praseodymium(3+)
165911
99.9%
7.07 g/mL

Chemical Identifiers

12036-32-7
329.812
MMKQUGHLEMYQSG-UHFFFAOYSA-N
165911
[O-2].[O-2].[O-2].[Pr+3].[Pr+3]
O3Pr2
MFCD00011178
dipraseodymium trioxide, praseodymium iii oxide, praseodymium oxide pr2o3, praseodynium oxide, dipraseodymium 3+ ion trioxidandiide, praseodymium iii oxide trace metals basis 10g
oxygen(2-);praseodymium(3+)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 234-857-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only