Learn More
Praseodymium(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
$230.10 - $938.66
Chemical Identifiers
CAS | 12036-32-7 |
---|---|
Molecular Formula | O3Pr2 |
Molecular Weight (g/mol) | 329.812 |
MDL Number | MFCD00011178 |
InChI Key | MMKQUGHLEMYQSG-UHFFFAOYSA-N |
Synonym | dipraseodymium trioxide, praseodymium iii oxide, praseodymium oxide pr2o3, praseodynium oxide, dipraseodymium 3+ ion trioxidandiide, praseodymium iii oxide trace metals basis 10g |
PubChem CID | 165911 |
IUPAC Name | oxygen(2-);praseodymium(3+) |
SMILES | [O-2].[O-2].[O-2].[Pr+3].[Pr+3] |
Description
Praseodymium(III) oxide is used as a pigment in glass, optic and ceramic industries. Praseodymium-doped glass called didymium glass is used in welding goggles, since it blocks infra-red radiation. Further, it is used as a dielectric in combination with silicon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12036-32-7 | |
329.812 | |
MMKQUGHLEMYQSG-UHFFFAOYSA-N | |
165911 | |
[O-2].[O-2].[O-2].[Pr+3].[Pr+3] |
O3Pr2 | |
MFCD00011178 | |
dipraseodymium trioxide, praseodymium iii oxide, praseodymium oxide pr2o3, praseodynium oxide, dipraseodymium 3+ ion trioxidandiide, praseodymium iii oxide trace metals basis 10g | |
oxygen(2-);praseodymium(3+) |
Specifications
2300°C | |
Sintered Lump | |
O3Pr2 | |
14,7712 | |
Insoluble in water. Moderately soluble in strong mineral acids. | |
[O-2].[O-2].[O-2].[Pr+3].[Pr+3] | |
329.812 | |
329.83 | |
Odorless | |
Praseodymium(III) oxide |
12036-32-7 | |
10 g | |
MFCD00011178 | |
dipraseodymium trioxide, praseodymium iii oxide, praseodymium oxide pr2o3, praseodynium oxide, dipraseodymium 3+ ion trioxidandiide, praseodymium iii oxide trace metals basis 10g | |
MMKQUGHLEMYQSG-UHFFFAOYSA-N | |
oxygen(2-);praseodymium(3+) | |
165911 | |
99.9% | |
7.07 g/mL | |
(metals basis) |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 234-857-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only