Contec™ PREempt™ RTU Disinfectant Solution
PREempt™ RTU utilizes AHP™, a patented blend when combined with low levels of hydrogen peroxide, increase efficacy and cleaning performance. When combined, it creates an ideal one step cleaner/disinfectant.
$130.37 - $165.24
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
29-636-6212
|
Contec™
11101 |
0.95 L |
Case of 12 Each for $130.37
|
|
|||||
29-636-6213
|
Contec™
11105 |
3.8 L |
Case of 4 Each for $165.24
|
|
|||||
Description
- AHP™ balances fast contact time while maintaining cleaning efficacy
- Carries no hazard classification
- PREempt™ RTU claims:
- Broad-spectrum sanitizing claim in 30 seconds
- Bactericidal and general virucide and tuberculocidal 3 minute kill claim
- Contains no volatile organic compounds
- Leaves minimal residues on surfaces once dry
- Ideal for cleaning and disinfecting environmental surfaces in cleanrooms and lab areas (work stations, fume hoods, equipment, hard non-porous environmental surfaces)
- Effective, fast-acting, one-step disinfectant cleaner
- Convenient, ready-to-use disinfectant for daily use
- Low residue and compatible with a wide range of materials
- Odor-free for good operator acceptability
- Does not require PPE
- Hydrogen peroxide breaks down into water and oxygen for easy disposal and minimal, to no environmental impact
- Not classified as dangerous goods
Specifications
Disinfectant Cleaner | |
Non-sterile |