Fischer Technical Company™ Prepared Culture Media in Plates
Prepared Culture Media in Plates labeled with product name, expiration date and lot number.
Supplier:  Fischer Technical Company™ CM145P25
 View more versions of this product
Catalog No. S716931A
Explore available promotions
Description
Description
  • Sterile plates are ready for immediate use
  • Supplied in disposable plastic petri dishes
  • Packed in clear, heat-sealed cellophane bags
Specifications
Specifications
2°C to 8°C
Plate
Nutrient Agar
Sterile
Microbial Identification
10 Plates/Pk.
15 mm
100 mm
Non-fastidious Microorganisms
US with Globally Sourced Materials
Prepared Media Plates
Bacteria
Microbiological Testing
0.59 in.
3.93 in.