Ergodyne™ ProFlex™ 100 Economy Spandex Back Support Brace
ProFlex™ 100 Economy Spandex Back Support is designed to remind workers of proper lifting techniques while also moving stress away from the spine for back pain relief.
Supplier:  Ergodyne™ 11388
 View more versions of this product
Catalog No. 19-819-461
Explore available promotions
Description
Description
  • Stretchable spandex body
  • Polypropylene stays for long-lasting integrity
  • Adjustable, detachable suspenders keep belt in place
  • Rubber track webbing prevents riding up
  • Machine wash cold, air dry
Specifications
Specifications
Black
Material Handling, Construction, Maintenance
25 to 58 in.
4X-Large
280D Spandex
64 to 147 cm