missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ergodyne™ ProFlex™ 100 Economy Spandex Back Support Brace

ProFlex™ 100 Economy Spandex Back Support is designed to remind workers of proper lifting techniques while also moving stress away from the spine for back pain relief.

$43.56 - $44.99

Specifications

Color Black
For Use With (Application) Material Handling, Construction, Maintenance
Material 280D Spandex
Size (English) Waist 25 to 58 in.
Size (Metric) Waist 64 to 147 cm
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-819-458
View Documents
Ergodyne™
11381
X-Small
Each for $43.56
 
19-819-460
View Documents
Ergodyne™
11387
3X-Large
Each for $44.99
 
19-819-461
View Documents
Ergodyne™
11388
4X-Large
Each for $44.99
 
Description

Description

  • Stretchable spandex body
  • Polypropylene stays for long-lasting integrity
  • Adjustable, detachable suspenders keep belt in place
  • Rubber track webbing prevents riding up
  • Machine wash cold, air dry
Specifications

Specifications

Black
280D Spandex
64 to 147 cm
Material Handling, Construction, Maintenance
25 to 58 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'