missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ergodyne™ ProFlex™ 1600 Standard Elastic Back Support Brace

ProFlex™ 1600 Standard Elastic Back Support is designed to remind workers of proper lifting techniques while moving stress away from the spine for back pain relief.

Supplier:  Ergodyne™ 11107

 View more versions of this product

Catalog No. 19-819-453


Explore available promotions

Description

Description

  • Premium 9 in. (22.9 cm) all-elastic body provides firm support
  • Contoured high-cut front panel for a comfortable fit
  • Polypropylene for long-lasting integrity
  • Adjustable, detachable suspenders keep belt in place
  • Rubber track webbing prevents riding up
  • Waist size ranges from 25 to 58 in. (64 to 147 cm)
  • Machine wash cold, air dry
Specifications

Specifications

Black
Material Handling, Construction,Warehousing/Distribution, Maintenance
9 in.
3X-Large
Suspenders
22.9 cm
Videos
SDS
Documents

Documents

Promotions

Promotions