missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega ADP-Glo™ Kinase Assay
ADP-Glo™ Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is converted into light by Ultra-Glo™ Luciferase. The luminescent signal positively correlates with kinase activity.
$1301.79 - $37045.64
Specifications
Content And Storage | -30°C to -10°C |
---|
Includes: ADP-Glo Reagent, Kinase Detection Buffer, Kinase Detection Substrate, Ultra Pure ATP (10mM), ADP (10mM)
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
PRV9103
![]() |
Promega
V9103 |
100,000 Assays |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
PRV9101
![]() |
Promega
V9101 |
1000 Assays |
Each for $1,301.79
|
|
||||||
PRV9102
![]() |
Promega
V9102 |
10,000 Assays |
Each for $7,428.63
|
|
||||||
PRV9104
![]() |
Promega
V9104 |
100,000 Assays, Bulk Pack |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
- One Assay for all your Kinase Screening Needs
- Use with virtually any kinase
- Stable luminescent signal
- Sensitive to low concentrations of ADP
Specifications
-30°C to -10°C |
- (a)Patents Pending.
- (b)Australian Pat. No. 2003268489 and other patents pending.
- (c)U.S. Pat. Nos. 6,602,677 and 7,241,584, Australian Pat. Nos. 754312 and 785294 and other patents and patents pending.
- (d)The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673.
- (e)Licensed from Lonza Nottingham Ltd. under U.S. Pat. Nos. 6,599,711 and 6,911,319 and other pending and issued patents.