Promega ADP-Glo™ Kinase Assay DFS Item
ADP-Glo™ Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is converted into light by Ultra-Glo™ Luciferase. The luminescent signal positively correlates with kinase activity.
Supplier:  Promega V9104
 View more versions of this product
Catalog No. PRV9104
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: ADP-Glo Reagent, Kinase Detection Buffer, Kinase Detection Substrate, Ultra Pure ATP (10mM), ADP (10mM)
Description
Description
  • One Assay for all your Kinase Screening Needs
  • Use with virtually any kinase
  • Stable luminescent signal
  • Sensitive to low concentrations of ADP
Specifications
Specifications
-30°C to -10°C
100,000 Assays, Bulk Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
  • (a)Patents Pending.
  • (b)Australian Pat. No. 2003268489 and other patents pending.
  • (c)U.S. Pat. Nos. 6,602,677 and 7,241,584, Australian Pat. Nos. 754312 and 785294 and other patents and patents pending.
  • (d)The method of recombinant expression of Coleoptera luciferase is covered by U.S. Pat. Nos. 5,583,024, 5,674,713 and 5,700,673.
  • (e)Licensed from Lonza Nottingham Ltd. under U.S. Pat. Nos. 6,599,711 and 6,911,319 and other pending and issued patents.