Promega Alkaline Phosphatase, Calf Intestinal DFS Item
Alkaline Phosphatase catalyzes the hydrolysis of 5'-phosphate groups from DNA, RNA and ribo- and deoxyribonucleoside triphosphates.
Supplier:  Promega M2825
 View more versions of this product
Catalog No. PR-M2825
Explore available promotions
Description
Description
  • Catalyzes the Hydrolysis of 5′ Phosphate Groups from DNA, RNA, and r- and dNTPs
  • Available at high concentration; Cat.# M2825 contains 1,000 units of CIAP at 20u/μL
  • Qualified for blue/white cloning; our assay provides a higher level of quality control for enzymes used in cloning applications
Specifications
Specifications
20 U/μL
10X Reaction Buffer: 0.5M Tris HCl (pH 9.3 at 25°C), 10mM MgCl2, 1mM ZnCl2, 10mM Spermidine
Prevent recircularization and religation of linearized cloning vector DNA by removing Phosphate groups from both 5'-termini, dephosphorylation of 5' Phosphorylated ends of DNA or RNA for subsequent labeling with (32P)ATP and T4 Polynucleotide Kinase
-30°C to -10°C
1000 U
Liquid
Colorless
8
10mM Tris HCl (pH 8), 1mM MgCl2, 0.1mM ZnCl2, 50mM KCl, 50% Glycerol
Calf intestinal mucosa
Alkaline Phosphatase