missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega AMV Reverse Transcriptase
AMV RT catalyzes DNA polymerization using template DNA, RNA or RNA:DNA hybrids.
$205.55 - $392.05
Specifications
Content And Storage | −30 to −10°C |
---|---|
Form | Liquid |
For Use With (Application) | cDNA Synthesis |
Source | Avian Myeloblastosis Virus |
Storage Buffer | Potassium Phosphate (pH 7.2 at °C) 200mM, 0.2% Triton™ X-100, DTT 2mM and Glycerol 50% |
Includes: 5X Reaction Buffer (250mM Tris-HCl [pH 8.3 at 25°C], 250mM KCl, 50mM MgCl2, 2.5mM spermidine, 50mM DTT)
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
PR-M5101
![]() |
Promega
M5101 |
AMV Reverse Transcriptase | 300U |
Each for $205.55
|
|
||||
PR-M9004
![]() |
Promega
M9004 |
AMV Reverse Transcriptase (HC) | 600U |
Each for $382.11
|
|
||||
PR-M5108
![]() |
Promega
PRM5108 |
AMV Reverse Transcriptase | 1000U |
Each for $392.05
|
|
||||
Description
- Efficient Reverse Transcription of Templates with High Secondary Structure
- DNA polymerization using template DNA, RNA or RNA:DNA hybrids
- Requires a primer as well as magnesium or manganese
- Stable at higher reaction temperatures (37-58°C)
- Supplied at a concentration of 10u/μL (Cat.# M5101, M5108) or 20-25u/μL (Cat.# M9004)
Specifications
−30 to −10°C | |
cDNA Synthesis | |
Potassium Phosphate (pH 7.2 at °C) 200mM, 0.2% Triton™ X-100, DTT 2mM and Glycerol 50% |
Liquid | |
Avian Myeloblastosis Virus |