missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega AMV Reverse Transcriptase DFS Item

AMV RT catalyzes DNA polymerization using template DNA, RNA or RNA:DNA hybrids.

Supplier:  Promega M9004

 View more versions of this product

Catalog No. PR-M9004


Explore available promotions

Includes: 5X Reaction Buffer (250mM Tris-HCl [pH 8.3 at 25°C], 250mM KCl, 50mM MgCl2, 2.5mM spermidine, 50mM DTT)

Description

Description

  • Efficient Reverse Transcription of Templates with High Secondary Structure
  • DNA polymerization using template DNA, RNA or RNA:DNA hybrids
  • Requires a primer as well as magnesium or manganese
  • Stable at higher reaction temperatures (37-58°C)
  • Supplied at a concentration of 10u/μL (Cat.# M5101, M5108) or 20-25u/μL (Cat.# M9004)
Specifications

Specifications

−30 to −10°C
Liquid
cDNA Synthesis
1 x 500μL 5X reaction buffer
Potassium Phosphate (pH 7.2 at °C) 200mM, 0.2% Triton™ X-100, DTT 2mM and Glycerol 50%
AMV Reverse Transcriptase (HC)
600U
20 to 25U/μL
Avian Myeloblastosis Virus
Videos
SDS
Documents

Documents

Promotions

Promotions