Promega BacTiter-Glo™ Microbial Cell Viability Assay Kit
The BacTiter-Glo™ Microbial Cell Viability Assay is a homogeneous method for determining the number of viable microbial cells in culture based on quantitation of the ATP present.
Supplier: Promega G8230
Includes: BacTiter-Glo Buffer; BacTiter-Glo Substrate (lyophilized)
Description
- A Luminescent Assay to Measure the Number of Viable Microbial Cells in Culture
- A homogeneous, single reagent assay
- Offers increased sensitivity, fast results
- U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
-30°C to -10°C | |
Microplate Reader | |
100 Assays at 100μL/assay in 96-well plates, 400 assays at 25μL/assay in 384-well plates |
Antimicrobial Compound Activity, Antimicrobial Compounds Screening, Viability Assay, Rapid Bacterial Quantitation, Rapid ATP Detection | |
10 mL |