Promega BacTiter-Glo™ Microbial Cell Viability Assay Kit
The BacTiter-Glo™ Microbial Cell Viability Assay is a homogeneous method for determining the number of viable microbial cells in culture based on quantitation of the ATP present.
$214.53 - $1138.45
Specifications
For Use With (Equipment) | Microplate Reader |
---|
Includes: BacTiter-Glo Buffer; BacTiter-Glo Substrate (lyophilized)
Catalog Number | Mfr. No. | For Use With (Application) | Quantity | Sufficient For | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Quantity | Sufficient For | Price | Quantity | |||
PR-G8231
|
Promega
G8231 |
Antimicrobial Compound Activity, Antimicrobial Compounds Screening, Viability Assay, Rapid Bacterial Quantitation, Rapid ATP Detection | 10 x 10mL | 1000 Assays at 100μL/assay in 96-well plates, 4000 assays at 25μL/assay in 384-well plates |
Each for $1,138.45
|
|
|||
PR-G8230
|
Promega
G8230 |
Antimicrobial Compound Activity, Antimicrobial Compounds Screening, Viability Assay, Rapid Bacterial Quantitation, Rapid ATP Detection | 10 mL | 100 Assays at 100μL/assay in 96-well plates, 400 assays at 25μL/assay in 384-well plates |
Each for $214.53
|
|
|||
PR-G8232
|
Promega
G8232 |
Antimicrobial Compound Activity, Antimicrobial Compounds Screening, Viability Assay, Rapid Bacterial Quantitation, Rapid ATP Detection | 100 mL | 1000 Assays at 100μL/assay in 96-well plates, 4000 assays at 25μL/assay in 384-well plates |
Each for $916.03
|
|
|||
Description
- A Luminescent Assay to Measure the Number of Viable Microbial Cells in Culture
- A homogeneous, single reagent assay
- Offers increased sensitivity, fast results
- U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Microplate Reader |