Promega BacTiter-Glo™ Microbial Cell Viability Assay Kit DFS Item
The BacTiter-Glo™ Microbial Cell Viability Assay is a homogeneous method for determining the number of viable microbial cells in culture based on quantitation of the ATP present.
Supplier:  Promega G8232
 View more versions of this product
Catalog No. PR-G8232
Explore available promotions
Includes: BacTiter-Glo Buffer; BacTiter-Glo Substrate (lyophilized)
Description
Description
  • A Luminescent Assay to Measure the Number of Viable Microbial Cells in Culture
  • A homogeneous, single reagent assay
  • Offers increased sensitivity, fast results
  • U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
-30°C to -10°C
Microplate Reader
1000 Assays at 100μL/assay in 96-well plates, 4000 assays at 25μL/assay in 384-well plates
Antimicrobial Compound Activity, Antimicrobial Compounds Screening, Viability Assay, Rapid Bacterial Quantitation, Rapid ATP Detection
100 mL