missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega CellTiter 96™ AQueous Nonradioactive Cell Proliferation Assay Kit (MTS)
Homogeneous, colorimetric cell viability assay; requires mixing of MTS and PMS components prior to assay, then a 1- to 4-hour incubation. Standalone MTS powder for applications where optimization of assay for specific cell types or conditions is desired.
$558.67 - $15953.51
Specifications
Detection Method | Colorimetric |
---|---|
For Use With (Application) | Determining the number of viable cells in proliferation |
Content And Storage | -30°C to -10°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR-G5421
|
Promega
G5421 |
1000 Assays |
Each for $558.67
|
|
|||||
PR-G5430
|
Promega
G5430 |
5000 Assays |
Each for $1,950.05
|
|
|||||
PRG5440
|
Promega
G5440 |
50,000 Assays |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- A Colorimetric Method for Determining the Number of Viable Cells
- Measures viable cells in proliferation, cytotoxicity or chemosensitivity assays
- Non-radioactive assay makes it safe and easy use
Order Info
CellTiter 96 AQueous MTS Reagent is also available separately in powdered form.
Specifications
Colorimetric | |
Determining the number of viable cells in proliferation | |
-30°C to -10°C |