Promega CellTiter 96™ AQueous Nonradioactive Cell Proliferation Assay Kit (MTS) DFS Item
Homogeneous, colorimetric cell viability assay; requires mixing of MTS and PMS components prior to assay, then a 1- to 4-hour incubation. Standalone MTS powder for applications where optimization of assay for specific cell types or conditions is desired.
Supplier:  Promega G5440
 View more versions of this product
Catalog No. PRG5440
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • A Colorimetric Method for Determining the Number of Viable Cells
  • Measures viable cells in proliferation, cytotoxicity or chemosensitivity assays
  • Non-radioactive assay makes it safe and easy use
Specifications
Specifications
Colorimetric
Determining the number of viable cells in proliferation
50,000 Assays
-30°C to -10°C