missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega CellTiter-Blue™ Cell Viability Assays DFS Item

Homogeneous, fluorometric cell viability assay (resazurin). Requires a 1- to 4-hour incubation.

$249.03 - $757.63

Specifications

Detection Method Fluorescence (preferred) or Absorbance
For Use With (Application) Monitoring Cell Viability
Format 96- or 384-well Formats
Content And Storage -30°C to -10°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity No. of Reactions Price Quantity  
Catalog Number Mfr. No. Quantity No. of Reactions Price Quantity  
PR-G8081 DFS Item
View Documents
Promega
G8081
100 mL 5000 assays in 96-well plates or 20,000 assays in 384-well plates
Each for $757.63
 
PR-G8080 DFS Item
View Documents
Promega
G8080
20 mL 1000 assays in 96-well plates or 4000 assays in 384-well plates
Each for $249.03
 
Description

Description

  • A Homogeneous, Fluorescent Assay to Monitor Cell Viability
  • Fast, flexible, time-saving
  • Amenable to multiplexing with other assays
Specifications

Specifications

Fluorescence (preferred) or Absorbance
Monitoring Cell Viability
96- or 384-well Formats
-30°C to -10°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'