Promega CellTiter-Blue™ Cell Viability Assays DFS Item
Homogeneous, fluorometric cell viability assay (resazurin). Requires a 1- to 4-hour incubation.
Supplier:  Promega G8081
 View more versions of this product
Catalog No. PR-G8081
Explore available promotions
Description
Description
  • A Homogeneous, Fluorescent Assay to Monitor Cell Viability
  • Fast, flexible, time-saving
  • Amenable to multiplexing with other assays
Specifications
Specifications
Fluorescence (preferred) or Absorbance
100 mL
-30°C to -10°C
Monitoring Cell Viability
96- or 384-well Formats
5000 assays in 96-well plates or 20,000 assays in 384-well plates