missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega CellTiter-Glo™ Luminescent Cell Viability Assay Kit
The most sensitive, fastest, homogeneous cell viability assay. Based on luminescent detection of ATP.
$222.68 - $7370.65
Specifications
For Use With (Application) | Determining the number of viable cells in culture based on the amount of ATP present |
---|---|
Format | 96- or 384-well Plates |
Content And Storage | -30°C to -10°C |
Includes: CellTiter-Glo Buffer; CellTiter-Glo Substrate (lyophilized)
Catalog Number | Mfr. No. | Quantity | No. of Reactions | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | No. of Reactions | Price | Quantity | |||||
PR-G7570
![]() |
Promega
G7570 |
10 mL | 100 x 100 μL or 400 x 25μL |
Each for $222.68
|
|
|||||
PR-G7571
![]() |
Promega
G7571 |
10 x 10 mL | 1000 x 100 μL or 4000 x 25 μL |
Each for $1,134.45
|
|
|||||
PR-G7572
![]() |
Promega
G7572 |
100 mL | 1000 x 100 μL or 4000 x 25 μL |
Each for $894.65
|
|
|||||
PR-G7573
![]() |
Promega
G7573 |
10 x 100 mL | 10,000 x 100 μL or 40,000 x 25μL |
Each for $7,370.65
|
|
|||||
Description
- A Luminescent Assay to Determine the Number of Viable Cells in Culture
- A homogeneous add-mix-measure format reduces the number of steps
- Higher level of detection requires fewer cells per assay
- Process plates consecutively with a very stable luminescent signal
- U.S. Pat. Nos. 6,602,677 and 7,241,584 and other patents and patents pending.
- U.S. Pat. No. 7,770,310 and other patents and patents pending.
- U.S. Pat. Nos. 7,083,911, 7,452,663 and 7,732,128 and other patents and patents pending.
- U.S. Pat. No. 7,741,067 and other patents and patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Determining the number of viable cells in culture based on the amount of ATP present | |
96- or 384-well Plates | |
-30°C to -10°C |